Skip to Content
Merck
All Photos(3)

Key Documents

D184101

Sigma-Aldrich

1,5-Dimethyl-2-pyrrolidinone

greener alternative

95%

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€133.00

€133.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
€133.00

About This Item

Empirical Formula (Hill Notation):
C6H11NO
CAS Number:
Molecular Weight:
113.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€133.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

form

liquid

greener alternative product score

old score: 5
new score: 4
Find out more about DOZN™ Scoring

greener alternative product characteristics

Design for Energy Efficiency
Reduce Derivatives
Real-Time Analysis for Pollution Prevention
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.465 (lit.)

bp

215-217 °C/743 mmHg (lit.)

density

0.982 g/mL at 25 °C (lit.)

greener alternative category

SMILES string

CC1CCC(=O)N1C

InChI

1S/C6H11NO/c1-5-3-4-6(8)7(5)2/h5H,3-4H2,1-2H3

InChI key

FILVIKOEJGORQS-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives. This product belongs to category of Re-engineered products, showing key improvements in Green Chemistry Principles “Reduce Derivatives”, “Real-Time Pollution Prevention” and “Design for Energy Efficiency”. Click here to view its DOZN scorecard.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

192.2 °F - closed cup

Flash Point(C)

89 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service