Assay
99%
refractive index
n20/D 1.542 (lit.)
bp
218-219 °C (lit.)
density
1.09 g/mL at 25 °C (lit.)
SMILES string
Cc1ccc2oc(C)nc2c1
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service