Skip to Content
Merck
All Photos(1)

Key Documents

D113204

Sigma-Aldrich

Dihydroxyfumaric acid hydrate

98%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€201.00

€201.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€201.00

About This Item

Empirical Formula (Hill Notation):
C4H4O6 · xH2O
CAS Number:
Molecular Weight:
148.07 (anhydrous basis)
Beilstein:
1724790
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€201.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

powder

mp

156 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

O.OC(=O)\C(O)=C(/O)C(O)=O

InChI

1S/C4H4O6.H2O/c5-1(3(7)8)2(6)4(9)10;/h5-6H,(H,7,8)(H,9,10);1H2/b2-1+;

InChI key

DDYHTXQJGPQZGN-TYYBGVCCSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vasudeva Naidu Sagi et al.
Journal of the American Chemical Society, 134(7), 3577-3589 (2012-01-28)
In the context of a "glyoxylate scenario" of primordial metabolism, the reactions of dihydroxyfumarate (DHF) with reactive small molecule aldehydes (e.g., glyoxylate, formaldehyde, glycolaldehyde, and glyceraldehyde) in water were investigated and shown to form dihydroxyacetone, tetrulose, and the two pentuloses

Questions

  1. How is this compound synthesized? I just want to know a synthetic route.

    1 answer
    1. This product is a Quality level 100. The methods of preparation are proprietary for products with Quality level 100. It may be helpful to view this publication: https://pubs.acs.org/doi/10.1021/ja01651a600.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service