Key Documents
CLS999075
Pyrex® ribbed watch glasses
O.D. ~75 mm
Synonym(s):
Corning® 9990
About This Item
UNSPSC Code:
41121500
NACRES:
NB.43
Pricing and availability is not currently available.
manufacturer/tradename
Corning 9990-75
O.D.
~75 mm
Related Categories
General description
Molded-in ribs allow vapors to escape during boiling to eliminate pressure buildup. Meets EPA requirements for methods 3005, 3010, 3020.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)