Key Documents
CLS893024
Pyrex® connecting distilling adapter
joint: ST/NS 24/40
Synonym(s):
Corning® 8930
About This Item
UNSPSC Code:
41121815
NACRES:
NB.43
Pricing and availability is not currently available.
Joint
joint: ST/NS24/40
manufacturer/tradename
Corning 8930-24
O.D.
22 mm
Related Categories
General description
The connecting arm is approximately at a 75° angle from vertical and is 200mm long.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)