Key Documents
CLS8180100
Pyrex® Lifetime Red graduated oil centrifuge tube
capacity 100.0 mL, O.D. × L 38 mm × 200 mm
Synonym(s):
Corning® 8180
About This Item
UNSPSC Code:
41121703
NACRES:
NB.43
Pricing and availability is not currently available.
material
conical bottom tube
packaging
case of 12
pack of 12
manufacturer/tradename
Corning 8180-100
O.D. × L
38 mm × 200 mm
capacity
100.0 mL
General description
In accordance with standard methods of testing for water and sediment in petroleum products. White graduations against a permanent red panel. Graduated upward to 0.5 mL in divisions of 0.05 mL; from 0.5 mL to 2 mL in divisions of 0.1 mL; from 2 mL to 3 mL in divisions of 0.2 mL; from 3 mL to 5 mL in divisions of 0.5 mL; from 5 mL to 10 mL in divisions of 1 mL; from 10 mL to 25 mL in divisions of 5 mL and at 50, 75 and 100 mL.
Legal Information
Corning is a registered trademark of Corning, Inc.
Lifetime-Red is a trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)