CLS56602L
Pyrex® Lifetime-Red™ volumetric flask with Pyrex® stopper
capacity 2,000 mL
Synonym(s):
flasks glass, volumetric flasks, volumetric glass, Corning® 5660
About This Item
Recommended Products
material
borosilicate glass
long
Joint
joint stopper27
sterility
non-sterile
feature
class A
closure type ST stopper
packaging
case of 4
manufacturer/tradename
Corning 5660-2L
tol. measuring range
0.50 mL
capacity
2,000 mL
Looking for similar products? Visit Product Comparison Guide
Features and Benefits
- Pyrex® Lifetime-Red™ volumetric flasks with Pyrex® stoppers (volume 2000 mL) have the capacity mark etched as a fine line through a narrow red band into clear glass, making a filler unnecessary and giving high legibility, accuracy and permanence.
- The strength of these flasks has been increased appreciably through machine-blown bodies to which are sealed heavy-tubing necks tooled for No. 27 Standard Taper glass stoppers.
- These Class A volumetric flasks have been manufactured to Class A tolerances as established by ASTM E- 694 for volumetric ware, ASTM E-542 for calibration of volumetric ware and ASTM E-288 for volumetric flasks. Calibrated.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service