Key Documents
CLS564650
Pyrex® Verified volumetric flasks
volume 50 mL
Synonym(s):
flasks glass, volumetric flasks, volumetric glass, Corning® 5646
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
borosilicate glass
long
sterility
non-sterile
feature
closure type ST stopper and Polyethylene stopper
packaging
case of 12
manufacturer/tradename
Corning 5646-50
capacity
50 mL
volume
50 mL
General description
Each individual flask′s measurement has been validated per ASTM E-542 during production, which guarantees conformance to Class A specifications called out in ASTM E-288. Each flask is labeled with "Verified" and a lot number. With this information, you can trace your glassware, access a Certificate of Analysis with lot specific data regarding both measurements and methods, and track usage.
All flasks are supplied with both a Pyrex stopper and HDPE stopper.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)