CLS55801
Pyrex® class A volumetric flask without snap cap
capacity 1 mL
Synonym(s):
clear flasks, pyrex flask, volumetric flasks, Corning® 5580
About This Item
Recommended Products
material
borosilicate glass
long
sterility
non-sterile
feature
cap: no
class A
packaging
case of 12
pack of 6
manufacturer/tradename
Corning 5580-1
tol. measuring range
0.01 mL
capacity
1 mL
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service