Key Documents
CLS49956L
Pyrex® wide-mouth graduated Erlenmeyer flask with screw-cap
capacity 6,000 mL
Synonym(s):
conical flasks, erlenmeyer flasks, flask erlenmeyer, flasks erlenmeyer, Corning® 4995
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
Pricing and availability is not currently available.
material
borosilicate glass
polypropylene cap
screw top cap
wide-mouth
sterility
non-sterile
feature
cap
closure type Screw cap
packaging
case of 1
manufacturer/tradename
Corning 4995-6L
capacity
6,000 mL
General description
Features a GL 45 size, PP, plug-seal screw-cap on a wide, 29 mm neck opening that accommodates larger volume serological pipettes. Also good for media prep and storage, or as a culture flask when optional membrane cap is used.
Other Notes
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)