CLS302510
Pyrex® double metric scale economy grade graduated cylinder
volume 10 mL
Synonym(s):
Corning® 3025
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
feature
subdivision 0.1 mL
manufacturer/tradename
Corning 3025-10
tol. measuring range
±5%
volume
10 mL
Looking for similar products? Visit Product Comparison Guide
General description
- Calibrated "to contain"
- double metric scale
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service