CLS213050
Pyrex® Precision-bore Class A burette
capacity 50 mL
Synonym(s):
burette precision, burettes, Corning® 2130
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass
feature
subdivision 0.10 mL
manufacturer/tradename
Corning 2130-50
tol. measuring range
0.05 mL
capacity
50 mL
Looking for similar products? Visit Product Comparison Guide
General description
With 2mm bore, stopcock, plug, and dustcover. Precision-bore tubing assures accuracy between any two points. Ground C glass stopcock gives precise control.
Other Notes
Class A burettes—Tolerances established by ASTM E 694 for volumetric ware, ASTM E 542 for calibration of volumetric ware, and ASTM E 287 for burettes.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service