Skip to Content
Merck
All Photos(1)

Key Documents

CF0017

Sigma-Aldrich

Imidazolyltetrafluorobromoethane

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€548.00

€548.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€548.00

About This Item

Linear Formula:
C5H3BrF4N2
CAS Number:
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22

€548.00


Check Cart for Availability

Request a Bulk Order

form

liquid

reaction suitability

reaction type: C-C Bond Formation

SMILES string

FC(F)(Br)C(F)(F)[n]1cncc1

InChI

1S/C5H3BrF4N2/c6-4(7,8)5(9,10)12-2-1-11-3-12/h1-3H

InChI key

WHWVFYIPMIWMMK-UHFFFAOYSA-N

Application

Imidazolyltetrafluorobromoethane is a fluoroalkylbromide that is a radical source of the imidazolyltetrafluoroethyl moiety. Alternatively, it can also be selectively metallated at the fluorinated carbon with Turbo Grignard reagent at low temperatures resulting in a thermally unstable anion that can act as a nucleophilic fluoroalkylation reagent towards a wide variety of electrophiles, such as aldehydes, ketones or sulfonylimines.

Automate your fluorination reactions with Synple Automated Synthesis Platform (SYNPLE-SC002)

Legal Information

Product of CF Plus Chemicals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Fluoroalkylation toolbox expands with various reagents for late-stage fluoroalkylation in organic synthesis and medicinal chemistry.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service