Skip to Content
Merck
All Photos(1)

Documents

CDS014072

Sigma-Aldrich

lysine acetylsalicylic acid

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H22N2O6
CAS Number:
Molecular Weight:
326.34
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:

form

solid

SMILES string

NCCCC[C@H](N)C(O)=O.CC(=O)Oc1ccccc1C(O)=O

InChI

1S/C9H8O4.C6H14N2O2/c1-6(10)13-8-5-3-2-4-7(8)9(11)12;7-4-2-1-3-5(8)6(9)10/h2-5H,1H3,(H,11,12);5H,1-4,7-8H2,(H,9,10)/t;5-/m.0/s1

InChI key

JJBCTCGUOQYZHK-ZSCHJXSPSA-N

Gene Information

Related Categories

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hassiba Chahdoura et al.
The Journal of pharmacy and pharmacology, 69(8), 1056-1063 (2017-04-28)
The aim of this study was to evaluate the analgesic and the anti-inflammatory activity of Opuntia microdasys at post flowering stage, F3 (OMF3) in rat and, in other hand, its antigenotoxic effects by the Allium cepa test. OMF3 extracts were
Mariangela Scavone et al.
Thrombosis and haemostasis, 120(10), 1442-1453 (2020-07-28)
Essential thrombocythemia (ET) patients are treated with aspirin (acetylsalicylic acid [ASA]) to prevent thrombosis. Previous studies showed that serum thromboxane (Tx) B2 was high 24 hours after enteric-coated (EC)-ASA in ET patients, due to increased number of noninhibited reticulated platelets (RPs)
Yosra Ayed et al.
Annals of clinical microbiology and antimicrobials, 11, 15-15 (2012-06-14)
Toxins derived from jellyfishes have been exploited as a model for the development of new drug promising applications to treat neurodegenerative diseases. The present work is aimed to evaluate the acute toxicity of crude venom of Pelagia noctiluca and then

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service