Skip to Content
Merck
All Photos(1)

Key Documents

CDS003997

Sigma-Aldrich

4-hydroxythiobenzamide

AldrichCPR

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€59.60

€59.60


Ships within 2 business days(Orders outside of US, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

Select a Size

Change View
250 MG
€59.60

About This Item

Empirical Formula (Hill Notation):
C7H7NOS
Molecular Weight:
153.20
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

€59.60


Ships within 2 business days(Orders outside of US, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

form

solid

SMILES string

NC(=S)c1ccc(O)cc1

InChI

1S/C7H7NOS/c8-7(10)5-1-3-6(9)4-2-5/h1-4,9H,(H2,8,10)

InChI key

VDTNKXSVUGXUOJ-UHFFFAOYSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mark Paul-Clark et al.
PloS one, 11(2), e0147289-e0147289 (2016-02-26)
Nonsteroidal anti-inflammatory drugs have been shown to reduce the incidence of gastrointestinal cancers, but the propensity of these drugs to cause ulcers and bleeding limits their use. H2S has been shown to be a powerful cytoprotective and anti-inflammatory substance in
Kuppusamy Sambathkumar
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 147, 51-66 (2015-04-02)
In this work, the experimental and theoretical study on molecular structure, vibrational spectral analysis of 4-hydroxythiobenzamide (HTB) have been reported. The solid phase FTIR (4000-400 cm(-1)) and FT-Raman spectra (3500-50 cm(-1)) were recorded. The molecular geometry, harmonic vibrational frequencies and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service