Skip to Content
Merck
All Photos(1)

Key Documents

CDS003526

Sigma-Aldrich

1-monobutyrin

AldrichCPR

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€85.60

€85.60


Ships within 2 business days(Orders outside of US, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

Select a Size

Change View
250 MG
€85.60

About This Item

Empirical Formula (Hill Notation):
C7H14O4
Molecular Weight:
162.18
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

€85.60


Ships within 2 business days(Orders outside of US, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

form

liquid

SMILES string

CCCC(=O)OCC(O)CO

InChI

1S/C7H14O4/c1-2-3-7(10)11-5-6(9)4-8/h6,8-9H,2-5H2,1H3

InChI key

RIEABXYBQSLTFR-UHFFFAOYSA-N

Related Categories

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhong-Biao Tan et al.
PloS one, 9(7), e102040-e102040 (2014-07-23)
A cDNA gene encoding a mature peptide of the mono- and diacylglycerol lipase (abbreviated to PcMdl) from Penicillium cyclopium PG37 was cloned and expressed in Pichia pastoris GS115. The recombinant PcMdl (rePcMdl) with an apparent molecular weight of 39 kDa
Matthew E R Butchbach et al.
Experimental neurology, 279, 13-26 (2016-02-20)
Proximal spinal muscular atrophy (SMA) is a childhood-onset degenerative disease resulting from the selective loss of motor neurons in the spinal cord. SMA is caused by the loss of SMN1 (survival motor neuron 1) but retention of SMN2. The number
A Bedford et al.
Poultry science, 97(4), 1315-1323 (2018-02-21)
Butyric acid is the primary energy source for colonocytes, and has shown potential as an alternative to in-feed antibiotics, due to its antimicrobial activity and positive effects on production performance traits of broiler chickens. SILOhealth 104 (SILO S.P.A., Florence, Italy)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service