Skip to Content
Merck
All Photos(1)

Key Documents

CDS003218

Sigma-Aldrich

2-ethylhexyl phosphate

AldrichCPR

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€85.60

€85.60


Ships within 2 business days(Orders outside of US, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

Select a Size

Change View
1 G
€85.60

About This Item

Empirical Formula (Hill Notation):
C8H19O4P
Molecular Weight:
210.21
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

€85.60


Ships within 2 business days(Orders outside of US, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

form

liquid

SMILES string

CCCCC(CC)COP(O)(O)=O

InChI

1S/C8H19O4P/c1-3-5-6-8(4-2)7-12-13(9,10)11/h8H,3-7H2,1-2H3,(H2,9,10,11)

InChI key

LJKDOMVGKKPJBH-UHFFFAOYSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

170.6 °F - closed cup

Flash Point(C)

77 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francisco Sánchez-Soberón et al.
Chemosphere, 209, 137-146 (2018-06-22)
In this pilot study three fractions of particulate matter (PM0.25, PM2.5-0.25, and PM10-2.5) were collected in three environments (classroom, home, and outdoors) in a village located nearby an industrial complex. Time-activity pattern of 20 students attending the classroom was obtained
Alba Maceira et al.
Chemosphere, 252, 126514-126514 (2020-03-23)
A multi-residue method based on gas chromatography-mass spectrometry combined with pressurised liquid extraction was developed to determine seven organophosphate esters (OPEs), six phthalate esters (PAEs), four benzotriazoles (BTRs), five benzothiazoles (BTHs) and four benzenesulfonamides (BSAs) in particulate matter samples from
Antonio Calvo-López et al.
Analytica chimica acta, 995, 77-84 (2017-11-12)
The construction and evaluation of a Cyclic Olefin Copolymer (COC)-based continuous flow potentiometric microanalyzer to simultaneously monitor potassium, chloride and nitrate ions in samples from an on-board water recycling process expected to be installed in future manned space missions is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service