Skip to Content
Merck
All Photos(1)

Key Documents

CDS000353

Sigma-Aldrich

1-Butylpyridinium chloride

AldrichCPR

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€71.00

€71.00


Ships within 2 business days(Orders outside of US, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

Select a Size

Change View
500 MG
€71.00

About This Item

Empirical Formula (Hill Notation):
C9H14ClN
Molecular Weight:
171.67
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

€71.00


Ships within 2 business days(Orders outside of US, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

form

solid

SMILES string

[Cl-].CCCC[n+]1ccccc1

InChI

1S/C9H14N.ClH/c1-2-3-7-10-8-5-4-6-9-10;/h4-6,8-9H,2-3,7H2,1H3;1H/q+1;/p-1

InChI key

POKOASTYJWUQJG-UHFFFAOYSA-M

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li Feng et al.
Journal of hazardous materials, 327, 11-17 (2016-12-30)
Traditional environmental risk assessment rarely focused on exposures to multi-component mixtures which may cause toxicological interactions and usually ignored that toxicity is a process in time, which may underestimate the environment risk of mixtures. In this paper, six chemicals belonging
Guifen Zhu et al.
Talanta, 200, 307-315 (2019-05-01)
A novel water compatible ciprofloxacin imprinted polymer is synthesized in water via a green, non-toxic and environmentally friendly polymerization process. Hydrophilic groups, including anionic chlorine, hydroxyl, and carbonyl oxygen provided by a bifunctional monomer comprising 1-allyl-3-vinylimidazole chloride and 2-hydroxyethyl methacrylate
Sagar Satpathi et al.
Physical chemistry chemical physics : PCCP, 18(43), 29740-29746 (2016-11-03)
Among different polymorphs of DNA, G-quadruplex (GQ) formation in guanine rich sequences has received special attention due to its direct relevance to cellular aging and abnormal cell growths. To date, smaller ions like Na
Guifen Zhu et al.
Journal of chromatography. A, 1592, 38-46 (2019-02-03)
Molecularly imprinted polymers (MIPs) synthesized by conventional functional monomers have poor specific recognition ability in strong polar solvent, which is not favorable to their applications in separation and analysis. In this work, an ionic liquid functional monomer, 1-allyl-3-vinylimidazolium chloridize, is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service