Skip to Content
Merck
All Photos(1)

Key Documents

CDPG-502

Sigma-Aldrich

Cationic PEGylated Liposomes

DOPC:Chol:DOTAP:DOPE-PEG(2000) (55:30:10:5 molar ratio 10mM)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352211
NACRES:
NA.25
Pricing and availability is not currently available.

Quality Level

composition

Phosphate buffered saline

concentration

10 mM

impurities

10 mol % DOTAP
30 mol % Cholesterol
5 mol % DOPE-PEG2000
55 mol % DOPC

particle size

100 nm

pH

7.4

Looking for similar products? Visit Product Comparison Guide

General description

Poly(ethylene glycol) (PEG) is commonly added to liposomes to improve stability, hydrophilicity and enhance circulation time. Anionic PEGylated liposomes protect liposomes from circulating proteins, improving their plasma clearance and enhancing their therapeutic effects. In addition, anionic liposomes demonstrate greater stability in solution, lead to lower aggregation when compared to neutral liposomes, and have increased endocytosis when compared with cationic and neutral liposomes.

Application

Drug delivery
Lipid-protein interactions

Storage and Stability

Liposomes should never be frozen. Liposomes should be stored in the dark at 4°C, except when brought to room temperature for brief periods prior to use.

Liposomes are made under sterile conditions. If you need to take multiple aliquots out of the vial, it is advised to take extreme care in not contaminating the vial. It is recommended to handle the vial under a sterile hood to maintain the sterility of the product. Liposomes should never be frozen. Ice crystals that form during freezing will rupture the lipid membrane of the liposomes and change the size of liposomes particles.

Legal Information

Cellsome is a trademark of Encapsula NanoSciences
Product of Encapsula Nanosciences

Disclaimer

For research use only

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service