Skip to Content
Merck
All Photos(3)

Key Documents

CAD7978

Micro-emulsifying needle

without reinforcing bar, gauge 20 ×, L 1 7/8 in.

Sign Into View Organizational & Contract Pricing

Select a Size

4 EA
€470.00

€470.00


Check Cart for Availability


Select a Size

Change View
4 EA
€470.00

About This Item

UNSPSC Code:
42142546
NACRES:
NB.03

€470.00


Check Cart for Availability

packaging

pack of 4 ea

manufacturer/tradename

Cadence Science 7978

L

1 7/8 in.

gauge

20 ×

Looking for similar products? Visit Product Comparison Guide

General description

Widely used for allergens and the mixing of adjuvants and antigens, oil based liquids or any two liquid media differing in viscosity. By locking two syringes on to the double hub needle or double hub adapter, two liquids such as a mineral oil and an emulsifying agent may be readily pumped between the two syringes to produce a uniform mixture.

Indicated lengths are the overall length. Researchers and technicians utilize Micro-Mate® 5 mL and 10 mL Luer lock tip glass syringes for micro-emulsifying procedures. The outstanding quality of the glass and the special secure Luer locking tip provide quality and reliability.

Legal Information

Micro-Mate is a registered trademark of Cadence Science

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service