Skip to Content
Merck
All Photos(1)

Key Documents

C70223

Sigma-Aldrich

4-Chloropyridine hydrochloride

99%

Synonym(s):

1-Nitro-4-iodobenzene, 4-Chloropyridine-HCl, 4-Chloropyridinium chloride, 4-Chloropyridinium monohydrochloride, 4-Iodo-1-nitrobenzene, 4-Nitro-1-iodobenzene, 4-Nitrophenyl iodide, p-Iodonitrobenzene, p-Nitroiodobenzene, p-Nitrophenyl iodide

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€79.60
100 G
€87.90

€79.60


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€79.60
100 G
€87.90

About This Item

Empirical Formula (Hill Notation):
C5H4ClN · HCl
CAS Number:
Molecular Weight:
150.01
Beilstein:
3684904
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€79.60


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

solid

mp

210 °C (subl.) (lit.)

SMILES string

Cl[H].Clc1ccncc1

InChI

1S/C5H4ClN.ClH/c6-5-1-3-7-4-2-5;/h1-4H;1H

InChI key

XGAFCCUNHIMIRV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

388.4 °F - closed cup

Flash Point(C)

198 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service