Skip to Content
Merck
All Photos(1)

Key Documents

C54902

Sigma-Aldrich

2-Chloromethyl-4-nitrophenol

96%

Synonym(s):

α-Chloro-4-nitro-o-cresol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2C6H3(NO2)OH
CAS Number:
Molecular Weight:
187.58
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

form

solid

mp

128 °C (dec.) (lit.)

SMILES string

Oc1ccc(cc1CCl)[N+]([O-])=O

InChI

1S/C7H6ClNO3/c8-4-5-3-6(9(11)12)1-2-7(5)10/h1-3,10H,4H2

InChI key

PJNPZIYGODMAQE-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service