Skip to Content
Merck
All Photos(1)

Key Documents

C117404

Sigma-Aldrich

Cyclopropanemethanol

98%

Synonym(s):

(Hydroxymethyl)cyclopropane, CPMO, Cyclopropyl carbinol, Cyclopropylmethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C3H5CH2OH
CAS Number:
Molecular Weight:
72.11
Beilstein:
1846846
MDL number:

Pricing and availability is not currently available.

Assay

98%

refractive index

n20/D 1.431 (lit.)

bp

123-124 °C/738 mmHg (lit.)

density

0.89 g/mL at 25 °C (lit.)

SMILES string

OCC1CC1

InChI

1S/C4H8O/c5-3-4-1-2-4/h4-5H,1-3H2

InChI key

GUDMZGLFZNLYEY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service