BR81939
BRAND® burette straight stopcock
for burette capacity 50 mL, PTFE key
Synonym(s):
Accessories, Burette stopcock, Burettes, Stopcocks
Select a Size
€49.70
Estimated to ship on17 April 2025
Select a Size
About This Item
€49.70
Estimated to ship on17 April 2025
Recommended Products
material
PTFE key
amber
packaging
pack of 1 ea
manufacturer/tradename
BRAND 81939
General description
Waiting times:
BLAUBRAND® - waiting time 30 sec. SILBERBRAND - no waiting time
A waiting time is usually not necessary if a titration is performed (see DIN EN ISO 385).
All BLAUBRAND® burettes are supplied with a batch number and an accompanying batch certificate in the original packaging.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)