Skip to Content
Merck
All Photos(1)

Documents

BR782090

BRAND® microplate BRANDplates®, cellGrade prem

size 96 wells, size 330 μL, flat bottom, white, with transparent bottom

Synonym(s):

BRANDplates®, Cell culture, Microplates, Tissue culture, cellGrade premium

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
NACRES:
NB.22

material

flat bottom
polystyrene
white wells

sterility

sterile

feature

lid
skirt (F-bottom)

packaging

pack of 50 ea
pkg of (individually packed)

manufacturer/tradename

BRAND 782090

size

330 μL
96 wells

working volume

330 μL

color

white, with transparent bottom

Looking for similar products? Visit Product Comparison Guide

General description

Poly-D-Lysine-equivalent surface, with analogous results regarding growth performance and cell morphology.Optimal adhesion of cells to the surface reduces cell damage when washing frequently.Cultivation of cell lines with the highest demands on their environment.Surface suited for serum-free and serum-reduced cultivation of cells.Good shelf life at room temperature.Alternative option to biologically coated surfaces.Clearly distinguishable through the color code: orange, embossed alphanumeric coding for 96-well standard plates.Free from endotoxins, DNase, DNA, RNase, non-cytotoxic.PS, sterile, with lids.BIO-CERT® CELL CULTURE QUALITY: Free of human DNA, RNase, DNase, Pyrogens and non cytotoxic

Features and Benefits

• Optimal surfaces for optimal results• Reliable quality from the cleanroom• Tested by independent laboratories

Legal Information

BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
BRANDplates is a registered trademark of BRAND GMBH + CO KG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service