Skip to Content
Merck
All Photos(1)

Key Documents

BR781982

BRAND® microplate BRANDplates®, cellGrade

size 384 wells, size 100 μL, flat bottom, black

Synonym(s):

BRANDplates®, Cell culture, Microplates, Tissue culture, cellGrade

Sign Into View Organizational & Contract Pricing

Select a Size

50 EA
€439.00

€439.00


Check Cart for Availability


Select a Size

Change View
50 EA
€439.00

About This Item

UNSPSC Code:
41122107
NACRES:
NB.22
material:
black wells
flat bottom
polystyrene
size:
100 μL
384 wells
sterility:
sterile
binding type:
cell culture treated
feature:
lid
skirt (F-bottom)

€439.00


Check Cart for Availability

material

black wells
flat bottom
polystyrene

sterility

sterile

feature

lid
skirt (F-bottom)

packaging

pack of 50 ea
pkg of (individually packed)

manufacturer/tradename

BRAND 781982

size

100 μL
384 wells

working volume

100 μL

color

black

binding type

cell culture treated

Looking for similar products? Visit Product Comparison Guide

General description

Standard plate for the cultivation of adherent cell lines.PS-surface with different chemical groups, such as carboxyl and hydroxyl groups, that are freely accessible.Surface is hydrophilic compared with non-treated PS.Serum components are easily bound onto the freely accessible chemical groups, allowing an indirect adhesion of cells.Clearly distinguishable through the color code: orange, embossed alphanumeric coding for 96-well standard plates.Free from endotoxins, DNase, DNA, RNase, non-cytotoxic.PS, sterile, with lids.BIO-CERT® CELL CULTURE QUALITY: Free of human DNA, RNase, DNase, Pyrogens and non cytotoxic

Features and Benefits

  • Optimal surfaces for optimal results
  • Reliable quality from the cleanroom
  • Tested by independent laboratories

Legal Information

BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
BRANDplates is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service