Skip to Content
Merck
All Photos(1)

Key Documents

BR780608

BRAND® 0.5 mL tube adapter for microcentrifuge tube rack 780605

Synonym(s):

Accessories, Microcentrifuge Tubes, Racks, Tubes

Sign Into View Organizational & Contract Pricing

Select a Size

20 EA
€13.00

€13.00


Check Cart for Availability


Select a Size

Change View
20 EA
€13.00

About This Item

UNSPSC Code:
41122804
NACRES:
NB.11

€13.00


Check Cart for Availability

material

polypropylene

packaging

pack of 20 ea

manufacturer/tradename

BRAND 780608

technique(s)

cryopreservation: suitable

General description

Microcentrifuge tube racks can be equipped with inserts to accommodate 0.5 mL microcentrifuge tubes. Easy permanent assembly.

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service