Key Documents
BR759242
BRAND® cap for UV-cuvette micro
green
Synonym(s):
Accessories, Caps, Cuvettes, UV-Cuvettes
Select a Size
100 EA
€14.20
€14.20
Check Cart for Availability
Select a Size
Change View
100 EA
€14.20
About This Item
UNSPSC Code:
41121800
NACRES:
SB.32
€14.20
Check Cart for Availability
material
PE
green
polyethylene
packaging
pack of 100 ea
manufacturer/tradename
BRAND 759242
General description
Round covers allow a tight closure and make it possible to store samples down to -20 °C.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)