Skip to Content
Merck
All Photos(1)

Key Documents

BR705874

BRAND® Transferpette® S Pipette

tip volume (10-100 μL), single-channel, adjustable volume, mechanical, DE-M

Synonym(s):

Air Displacement Pipette, Micropipette, Micropipettor, Pipette, Pipettor, Transferpette S, microliter pipette, microliter pipettor

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
€270.00

€270.00


Check Cart for Availability


Select a Size

Change View
1 EACH
€270.00

About This Item

UNSPSC Code:
41121600
NACRES:
NB.01

€270.00


Check Cart for Availability

feature

single-channel adjustable volume
mechanical

packaging

case of 1 ea

manufacturer/tradename

BRAND 705874

tip volume

(10-100 μL)

Looking for similar products? Visit Product Comparison Guide

General description

The Transferpette® S air displacement pipette allows you to work efficiently and ergonomically with both small and large volumes. Low operating forces and a short stroke ensure relaxed pipetting, even during long pipetting series. Due to its perfect ergonomics, the Transferpette S provides a comfortable grip in any hand position, whether you are right- or left-handed, or have big or small hands. Changing your grip is not necessary, since the microliter pipette provides true one-hand operation: set the volume, pipette, eject the tip. The volume-change protection is also easy to operate with gloves on and protects your set volume against accidental adjustment. The Transferpette S also features Easy Calibration technology, which makes calibration a breeze. Calibration can be carried out without tools. A compatibility table shows which tips fit which device, so that you always order the correct pipette tips.

Application

Pipetting

Features and Benefits

  • Efficient: one-hand operation
  • Flexible: ergonomic design for all hand sizes
  • Reliable: volume-change protection prevents accidental adjustments


Limitations of use:
  • Operating temperature of instrument and reagent should be between +15 °C and +40 °C (59 °F to 104 °F)
  • Vapor pressure up to 500 mbar
  • Viscosity: 260 mPa s

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG
Transferpette is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service