Skip to Content
Merck
All Photos(1)

Key Documents

BR704006

BRAND® bottle for dispensers

capacity 500 mL, with Ethyl-Acrylate coat

Synonym(s):

Accessories, Bottle-top Dispenser, Bottles, Dispensette®

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€13.00

€13.00


Check Cart for Availability


Select a Size

Change View
1 EA
€13.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.43

€13.00


Check Cart for Availability

material

amber bottle
narrow-mouth bottle
polypropylene cap
soda-lime glass bottle (with Ethyl-Acrylate coat)
square bottle

Joint

Joints threaded neck (GL32)

feature

with cap
with Ethyl-Acrylate coat
with PP screw cap GL32

packaging

pack of 1 ea

manufacturer/tradename

BRAND 704006

capacity

500 mL

diam. × H

80 mm × 195 mm

Looking for similar products? Visit Product Comparison Guide

General description

Space-saving square base. Available ethylene-acrylate-coated or uncoated. The coating envelops the glass bottle like a protection skin. Should the bottle break, the splintering hazards are reduced considerably. The maximum operating temperature for coated bottles is 80 °C. To preserve the coating, do not clean at temperatures exceeding 60 ºC.

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service