Key Documents
BR37241
BRAND® volumetric flask BLAUBRAND® class A DE-M marking
capacity 10 mL, joint: 7/16, polypropylene stopper
Select a Size
2 EA
€36.40
€36.40
Estimated to ship on13 April 2025
Select a Size
Change View
2 EA
€36.40
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
€36.40
Estimated to ship on13 April 2025
material
borosilicate glass 3.3 flask
polypropylene stopper
Joint
joint ST/NS neck7/16
joint: 7/16
feature
class A DE-M marking. DIN EN ISO 1042. Calibrated to contain (TC, In).
packaging
pack of 2 ea
manufacturer/tradename
BRAND 37241
capacity
10 mL
General description
The BRAND® volumetric flask BLAUBRAND® class A with DE-M marking is made from high-quality Boro 3.3 glass and complies with DIN EN ISO 1042 standards. It is calibrated to contain (TC, In), ensuring accurate liquid measurement for various laboratory applications. Each volumetric flask is delivered with a batch number and batch certificate included in the original packaging, providing quality assurance and traceability. This reliable volumetric flask is an essential tool for precise analytical work in any laboratory setting.
Legal Information
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)