BR114106
BRAND® culture tube, rimless
L × W 12 mm × 75 mm, thickness 0.6 mm
Synonym(s):
Culture tubes, Tubes
All Photos(1)
About This Item
Recommended Products
material
round bottom
screw top cap
soda-lime glass tube
straight neck
transparent tube
sterility
non-sterile
feature
RCF 3,000 × g
neck thread push fit
packaging
pack of 250 ea
manufacturer/tradename
BRAND 114106
L × W
12 mm × 75 mm
capacity
12 mL
5.5 mL
thickness
0.6 mm
General description
Crafted from high-quality glass, these tubes provide excellent clarity and durability, making them ideal for cell culture and other scientific applications. Autoclavable at 121 °C, ensuring easy sterilization.
Features and Benefits
• Excellent sample visibility• Good chemical resistance
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG