BR113125
BRAND® forcep with rectangular ends
PTFE, L 100 mm
Synonym(s):
Forceps, PTFE
All Photos(1)
About This Item
Recommended Products
material
PTFE
manufacturer/tradename
BRAND 113125
L
100 mm
Related Categories
General description
The BRAND® Forceps with Rectangular Ends are specialized tools designed for handling materials in challenging laboratory environments. Made from high-quality PTFE (polytetrafluoroethylene), these forceps offer exceptional durability and performance. The PTFE construction ensures excellent resistance to a wide range of chemicals, making these forceps suitable for use in corrosive environments. Capable of withstanding high temperatures, these forceps are ideal for applications that involve heat exposure.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG