Key Documents
BMSH6004
Sigma-Aldrich
Incu-Mixer MP4 Heated 4 Plate Vortexer
AC/DC input 115 V AC, US 3-pin plug
Synonym(s):
incubated microplate stirrer, microwellplate vortexer, multiplate incubator
About This Item
UNSPSC Code:
41103800
NACRES:
NB.77
Pricing and availability is not currently available.
packaging
pack of 1 ea
manufacturer/tradename
Benchmark Scientific H6004
parameter
100 rpm speed (1500 rpm)
5 - 70 °C temp. range
W × D × H
12.6 in. × 13.8 in. × 7.3 in.
size
, no of position=4
General description
The new Incu-Mixer MP series is designed to provide an optimal method for vortexing and incubating microplates. Simultaneous heating from both the bottom surface and the lid provide temperature uniformity throughout the sample. Additionally, the heated lid minimizes evaporation and prevents condensation.
A compact, 2mm orbit ensures thorough mixing in microplates without splashing. Speed, time and temperature are all displayed in real time on the large LCD screen while an internal validation system constantly monitors all parameters for accuracy.
Designed for application flexibility, the Incu-Mixer series is available in two, and four-plate capacities, both capable of accepting all standard plates up to 40mm high. Independent time, temperature and speed controls allow the mixer to function as 3 machines in one; a multi-plate incubator, vortexer, or a combination of the two. The Incu-Mixer MP series is ideal for a wide variety of molecular biology applications including immunochemical reactions, enzyme and protein analysis, and microarray analysis.
  • Optimized for incubated mixing in microplates
  • Heated lid prevents condensation
  • Independently control time, temp. and speed
Choose from one of the most recent versions:
Certificates of Analysis (COA)