Skip to Content
Merck
All Photos(1)

Key Documents

BML00172

Sigma-Aldrich

3,4-diaminophenylboronic acid, pinacol ester

Synonym(s):

4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene-1,2-diamine

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€556.00

€556.00


Ships within 2 business days(Orders outside of US, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

Select a Size

Change View
1 G
€556.00

About This Item

Empirical Formula (Hill Notation):
C12H19BN2O2
CAS Number:
Molecular Weight:
234.10
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

€556.00


Ships within 2 business days(Orders outside of US, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

form

solid

SMILES string

CC(C(C)(C)O1)(C)OB1C2=CC=C(N)C(N)=C2

InChI

1S/C12H19BN2O2/c1-11(2)12(3,4)17-13(16-11)8-5-6-9(14)10(15)7-8/h5-7H,14-15H2,1-4H3

InChI key

SVUDHDDKOKWBNK-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service