Skip to Content
Merck
All Photos(1)

Key Documents

BASMF2062

Sigma-Aldrich

Non-Aqueous Reference Electrode Kit

Synonym(s):

Kit for Non-Aqueous Reference Electrode

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
€333.00

€333.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 KIT
€333.00

About This Item

UNSPSC Code:
12352300
NACRES:
NA.23

€333.00


Check Cart for Availability

Request a Bulk Order

General description

Kit includes:
  • Non-Aqueous Reference Electrode (1) - BASMF1085
  • 1g Silver Nitrate (1)
  • 1/8" chunks CoralPor with 1/2" tubes PTFE heat shrink (1/pk) - BASMF2064
  • Reference Electrode O-ring (1)
Non-Aqueous Reference Electrode Kit- Ag/Ag+

Legal Information

Product of Bioanalytical Systems, Inc.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Ox. Sol. 2 - Skin Corr. 1B

Storage Class Code

5.1B - Oxidizing hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service