Key Documents
BAH369401010
SP Bel-Art® Sterileware Long Handled Sampling Spoon
white polystyrene, nominal capacity 1.25 mL , ¼tsp, L × W 17.8 cm (7 in.) × 0.8125 in. (20 mm), sterile, pack of 10 ea (double bagged)
Synonym(s):
Lab Scoop, dispensing tool, spoon
About This Item
UNSPSC Code:
41122800
NACRES:
NB.35
Pricing and availability is not currently available.
material
white polystyrene
sterility
sterile
packaging
pack of 10 ea (double bagged)
manufacturer/tradename
Bel-Art H36940-1010
L × W
17.8 cm (7 in.) × 0.8125 in. (20 mm)
nominal capacity
1.25 mL , ¼tsp
General description
The long handle on these spoons enables users to reach into jars, bottles and other confined areas to minimize the potential of personal contact with sampled material.
Features and Benefits
  • Ten individually packaged spoons are sealed in an outer bag to create multiples layers of protection against contaminants and debris
  • Ideal for use in multi-zone clean rooms
  • FDA grade polystyrene safe for food, drug and cosmetic use
  • Gamma irradiated sterile to SAL 10-6
  • Lot stamped for accurate lot tracing
Note: Sterilized products are not returnable.
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Sterileware is a trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)