BAF249800015
Micro-centrifuge rack box for β-emitters
W × L × H 20 cm (8 in.) × 9 cm (3.5 in.) × 6 cm (2.25 in.)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
material
acrylic
feature
Holds 36 x 1.5 mL tubes
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F24980-0015
W × L × H
20 cm (8 in.) × 9 cm (3.5 in.) × 6 cm (2.25 in.)
Related Categories
General description
Box is fabricated from 9.5 mm (3/8 in.) thick acrylic and has tight fitting cover to maximize protection. Non-skid feet provide stability.
- Acrylic storage box contains beta radiation to protect surroundings
- Holds 36 x 1.5 mL microcentrifuge tubes
- Water or crushed ice can be placed under rack
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service