Skip to Content
Merck
All Photos(1)

Key Documents

BAF131741012

Cleanware self-adhesive waste bags

W × L 305 mm (12 in.) × 406 mm (16 in.)

Sign Into View Organizational & Contract Pricing

Select a Size

50 EA
€48.60

€48.60


Check Cart for Availability


Select a Size

Change View
50 EA
€48.60

About This Item

UNSPSC Code:
24111500
NACRES:
NB.35

€48.60


Check Cart for Availability

packaging

pack of 50 ea

manufacturer/tradename

Bel-Art F13174-1012

W × L

305 mm (12 in.) × 406 mm (16 in.)

Looking for similar products? Visit Product Comparison Guide

General description

These disposable waste bags feature an adhesive strip across the top so they can be hung from walls, cabinets, or virtually anywhere freeing bench or hood space. The adhesive backing strip is easily peeled off, and the bag can then be hung where needed. The back panel is longer than the front for easy opening. Maximum weight of contents should not exceed 1.36 kg (3 lb.). Packed 50 bags per pack
  • Convenient, hang-anywhere design
  • Simply peel off the backing and hang close to your work area
  • Made of High Molecular Weight High Density (HMHD) Polyethylene
  • Steam autoclavable at 121 ºC (250 ºF)

Legal Information


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service