All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C8H8N4O
CAS Number:
Molecular Weight:
176.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
98%
form
powder
mp
>300 °C (lit.)
SMILES string
NC(=O)Nc1nc2ccccc2[nH]1
InChI
1S/C8H8N4O/c9-7(13)12-8-10-5-3-1-2-4-6(5)11-8/h1-4H,(H4,9,10,11,12,13)
InChI key
LNYZEFQMIURYEI-UHFFFAOYSA-N
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
S Trivulzio
Pharmacological research communications, 20(12), 1035-1046 (1988-12-01)
2-Benzimidazolylurea (BIU) decreases gastric acid secretion. The antisecretive activity appears to be associated with antihistaminic and antimuscarinic effects. The antihistaminic activity of BIU appears from its inhibitory effects on betazole stimulated gastric acid secretion and from its inhibitory activity on
R S Gupta
Biochemical and biophysical research communications, 143(1), 225-232 (1987-02-27)
The cross resistance patterns of a set of nocodazole-resistant (NocR) and podophyllotoxin-resistant (PodR) mutants of Chinese hamster ovary cells, which exhibit highly specific cross resistance towards compounds that show nocodazole-like antimitotic activity, towards a large number of benzimidazole derivatives have
Liang Hong et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(27), 9971-9976 (2014-06-10)
The voltage-gated proton channel Hv1 plays important roles in proton extrusion, pH homeostasis, and production of reactive oxygen species in a variety of cell types. Excessive Hv1 activity increases proliferation and invasiveness in cancer cells and worsens brain damage in
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service