Skip to Content
Merck
All Photos(1)

Key Documents

B53383

Sigma-Aldrich

Bis(tributyltin) oxide

96%

Synonym(s):

Bis[tri-n-butyltin(IV)]oxide, HBD, Hexabutyldistannoxane, TBTO, Tributyltin(IV) oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)3SnOSn(CH2CH2CH2CH3)3
CAS Number:
Molecular Weight:
596.10
Beilstein:
745057
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

<0.01 mmHg ( 25 °C)

Assay

96%

form

liquid

refractive index

n20/D 1.486 (lit.)

bp

180 °C/2 mmHg (lit.)

density

1.17 g/mL at 25 °C (lit.)

SMILES string

CCCC[Sn](CCCC)(CCCC)O[Sn](CCCC)(CCCC)CCCC

InChI

1S/6C4H9.O.2Sn/c6*1-3-4-2;;;/h6*1,3-4H2,2H3;;;

InChI key

APQHKWPGGHMYKJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Employed in the synthesis of α,β-unsaturated methyl ketones,[1] isoxazoles,[2] fungicides,[3] and bactericides.[4]

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT RE 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Chemistry Letters (Jpn), 1219-1219 (1993)
Applied Organometallic Chemistry, 8, 37-37 (1994)
Journal of the Chemical Society. Perkin Transactions 1, 413-413 (1994)
J. Polym. Mater., 9, 297-297 (1992)
Ahmed M Osman et al.
Toxicological sciences : an official journal of the Society of Toxicology, 126(1), 84-100 (2011-12-17)
We report the results of phosphoproteomic analysis of mouse thymoma cells treated with tributyltin oxide (TBTO), an immunotoxic compound. After cell lysis, phosphoproteins were isolated using Phosphoprotein Purification Kit, separated by SDS-PAGE and subsequently digested with trypsin. Phosphopeptides were enriched

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service