B48207
Bis(2-methoxyethyl)amine
98%
Synonym(s):
2,2′-Dimethoxydiethylamine, 2-Methoxy-N-(2-methoxyethyl)ethanamine, Di(2-methoxyethyl)amine, N,N-Bis(2-methoxyethyl)amine
Select a Size
About This Item
Recommended Products
Assay
98%
form
liquid
refractive index
n20/D 1.419 (lit.)
bp
170-172 °C (lit.)
density
0.902 g/mL at 25 °C (lit.)
SMILES string
COCCNCCOC
InChI
1S/C6H15NO2/c1-8-5-3-7-4-6-9-2/h7H,3-6H2,1-2H3
InChI key
IBZKBSXREAQDTO-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
- In the Pd-catalyzed Buchwald-Hartwig amination reaction to synthesize aryl amines by reacting with aryl and heteroaryl halides.
- To synthesize α-ketoamides via metal-free oxidative coupling reaction with methyl ketones in the presence of tert-butyl hydroperoxide as an oxidant and iodine as a catalyst.
- To synthesize N, N-dialkyl perfluoroalkanesulfonamides by reacting with perfluoroalkanesulfonyl halides.
- To synthesize N-(alkoxy)succinamide ester by reacting with succinyl chloride, which can be used as an intermediate to prepare diol monomer for biodegradable polyester synthesis.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
136.4 °F - closed cup
Flash Point(C)
58 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service