Skip to Content
Merck
All Photos(1)

Documents

B27005

Sigma-Aldrich

3-Benzyloxybenzaldehyde

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2OC6H4CHO
CAS Number:
Molecular Weight:
212.24
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

chunks

mp

56-58 °C (lit.)

SMILES string

O=Cc1cccc(OCc2ccccc2)c1

InChI

1S/C14H12O2/c15-10-13-7-4-8-14(9-13)16-11-12-5-2-1-3-6-12/h1-10H,11H2

InChI key

JAICGBJIBWDEIZ-UHFFFAOYSA-N

Application

Some of the reported applications of 3-benzyloxybenzaldehyde are:
  • Synthesis of silybin analogs as anticancer agents that produce apoptosis in ovarian cancer cells through tubulin inhibition.
  • Synthesis of porphyrin and boron dipyrromethene (BODIPY) derivatives for fluorescent applications.
  • Preparation of styrene copolymers with varying chain mobilities.
  • Synthesis of aromatic derivatives of trimethoprim as potential antimalarial agents.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Design and discovery of silybin analogues as antiproliferative compounds using a ring disjunctive?Based, natural product lead optimization approach.
Manivannan E, et al.
European Journal of Medicinal Chemistry, 133, 365-378 (2017)
Novel Copolymers of Styrene. 2. Oxy Ring-Substituted 2-Cyano-3-phenyl 2-propenamides.
Kharas G B, et al.
Journal of Macromolecular Science, Part A: Pure and Applied Chemistry, 50(8), 797-802 (2013)
Target guided synthesis of 5-benzyl-2, 4-diamonopyrimidines: their antimalarial activities and binding affinities to wild type and mutant dihydrofolate reductases from Plasmodium falciparum.
Sirichaiwat C, et al.
Journal of Medicinal Chemistry, 47(2), 345-354 (2004)
Synthesis of a novel BODIPY library and its application in the discovery of a fructose sensor.
Zhai D, et al.
ACS Combinatorial Science, 14(2), 81-84 (2012)
Synthesis of Functionalized trans?A2B2?Porphyrins Using Donor?Acceptor Cyclopropane?Derived Dipyrromethanes.
Beyzavi M H, et al.
Advanced Synthesis & Catalysis, 355(7), 1409-1422 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service