Skip to Content
Merck
All Photos(1)

Key Documents

B104256

Sigma-Aldrich

Butyrylthiocholine iodide

98%

Synonym(s):

(2-Mercaptoethyl)trimethylammonium iodide butyrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3N(I)CH2CH2SCOCH2CH2CH3
CAS Number:
Molecular Weight:
317.23
Beilstein:
3729509
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

98%

mp

171-174 °C (lit.)

SMILES string

[I-].CCCC(=O)SCC[N+](C)(C)C

InChI

1S/C9H20NOS.HI/c1-5-6-9(11)12-8-7-10(2,3)4;/h5-8H2,1-4H3;1H/q+1;/p-1

InChI key

WEQAAFZDJROSBF-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Denis Josse et al.
Chemico-biological interactions, 232, 94-100 (2015-03-21)
In this work, our goals were to establish whether hair decontamination by showering one hour post-exposure to the highly toxic organophosphate nerve agent VX was effective, whether it required the addition of a detergent to water and, if it could
Cristobal Narvaez et al.
Chemosphere, 135, 75-82 (2015-04-29)
Inhibition of blood esterase activities by organophosphate (OP) pesticides has been used as a sensitive biomarker in birds. Furthermore, compared to mammalian vertebrates, less is known about the role of these enzyme activities in the digestive tracts of non-mammalian vertebrates
Gabriele Horn et al.
Archives of toxicology, 89(3), 405-414 (2014-06-11)
Organophosphorus compounds (OP) are bound to human butyrylcholinesterase (BChE) and endogenous or exogenous BChE may act as a stoichiometric scavenger. Adequate amounts of BChE are required to minimize toxic OP effects. Simultaneous administration of BChE and oximes may transfer the
Adriana Ferreira Lopes Vilela et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 968, 87-93 (2013-12-11)
The discovery of selective inhibitors for acetylcholinesterase (AChE) or butyrylcholinesterase (BChE) is extremely important for the development of drugs that can be used in the treatment of patients diagnosed with the Alzheimer's disease (AD). For this reason, there is a
Stefan Schwarz et al.
Bioorganic & medicinal chemistry, 22(13), 3370-3378 (2014-05-24)
The development of remedies against the Alzheimer's disease (AD) is one of the biggest challenges in medicinal chemistry nowadays. Although not completely understood, there are several strategies fighting this disease or at least bringing some relief. During the progress of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service