storage temp.
2-8°C
SMILES string
CC(CCC)(C)NC1=CC=C(NC(C)=O)C=C1
InChI
1S/C14H22N2O/c1-5-10-14(3,4)16-13-8-6-12(7-9-13)15-11(2)17/h6-9,16H,5,10H2,1-4H3,(H,15,17)
InChI key
WMTCHYABBOFZTB-UHFFFAOYSA-N
Application
related product
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service