Skip to Content
Merck
All Photos(1)

Key Documents

ALD00619

Sigma-Aldrich

1,4-Diethyl 2-(1-pyridin-1-ium-yl)butanedioate ethylsulfate

≥95%

Synonym(s):

Fumarate-Pyr

Sign Into View Organizational & Contract Pricing

Select a Size

2 G
€317.00

€317.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
2 G
€317.00

About This Item

Empirical Formula (Hill Notation):
C15H23NO8S
Molecular Weight:
377.41
UNSPSC Code:
12352100
NACRES:
NA.22

€317.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥95%

form

powder and chunks

mp

65-70 °C

Application

1,4-Diethyl 2-(1-pyridin-1-ium-yl)butanedioate ethylsulfate is a bench-stable pyridinium feedstock capable of acting as an inexpensive, scalable and practical building block toward the synthesis of a variety of C-4 alkyl pyridines.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jin Choi et al.
Journal of the American Chemical Society, 143(31), 11927-11933 (2021-07-29)
The direct position-selective C-4 alkylation of pyridines has been a long-standing challenge in heterocyclic chemistry, particularly from pyridine itself. Historically this has been addressed using prefunctionalized materials to avoid overalkylation and mixtures of regioisomers. This study reports the invention of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service