Skip to Content
Merck
All Photos(1)

Key Documents

ALD00460

Sigma-Aldrich

Sodium 2-(4-Bromophenyl)-1,1-difluoroethanesulfinate

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€267.00

€267.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
€267.00

About This Item

Empirical Formula (Hill Notation):
C8H6BrF2NaO2S
Molecular Weight:
307.09
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

€267.00


Check Cart for Availability

Request a Bulk Order

form

powder

Quality Level

reaction suitability

reaction type: C-C Bond Formation

mp

275 °C

SMILES string

FC(F)(S(O[Na])=O)CC1=CC=C(Br)C=C1

InChI

1S/C8H7BrF2O2S.Na/c9-7-3-1-6(2-4-7)5-8(10,11)14(12)13;/h1-4H,5H2,(H,12,13);/q;+1/p-1

InChI key

MGXWINRJXWSLAL-UHFFFAOYSA-M

Application

The following Baran Sulfinate is a part of a toolbox of diversification reagents, which are ideal for the late-stage functionalization of nitrogen-containing heterocycles. The regioselectivity can be effectively tuned by modification of the pH and solvent selection.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service