Skip to Content
Merck
All Photos(1)

Key Documents

ALD00430

Sigma-Aldrich

1-Naphthalenyl ester fluorosulfuric acid

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€11.70

€11.70


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
€11.70

About This Item

Empirical Formula (Hill Notation):
C10H7FO3S
CAS Number:
Molecular Weight:
226.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€11.70


Check Cart for Availability

Request a Bulk Order

description

Fp: >230°F

Quality Level

form

liquid

refractive index

n20/D 1.5576

density

1.3674 g/mL at 25 °C

SMILES string

O=S(OC1=CC=CC2=CC=CC=C21)(F)=O

InChI

1S/C10H7FO3S/c11-15(12,13)14-10-7-3-5-8-4-1-2-6-9(8)10/h1-7H

InChI key

GEZWNEDYJCOWLU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

The following aryl fluorosulfate can be utilized as a cross-coupling partner in Suzuki-Miyaura reactions. The standard reaction conditions for these palladium-catalyzed C-C bond formations are ligand-free and can be performed in water, at room temperature and are not air sensitive.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service