Skip to Content
Merck
All Photos(1)

Key Documents

ALD00382

Sigma-Aldrich

1,5-Bis[4-(trifluoromethyl)phenyl]-1,4-pentadien-3-one

Synonym(s):

(1E,4E)-1,5-Bis[4-(trifluoromethyl)phenyl]1,4-pentadien-3-on, CF3-dba

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€11.80

€11.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
€11.80

About This Item

Empirical Formula (Hill Notation):
C19H12F6O
CAS Number:
Molecular Weight:
370.29
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€11.80


Check Cart for Availability

Request a Bulk Order

form

solid

Quality Level

reaction suitability

reagent type: catalyst
reagent type: ligand
reaction type: C-H Activation

storage temp.

2-8°C

SMILES string

O=C(/C=C/C1=CC=C(C(F)(F)F)C=C1)/C=C/C2=CC=C(C(F)(F)F)C=C2

InChI

1S/C19H12F6O/c20-18(21,22)15-7-1-13(2-8-15)5-11-17(26)12-6-14-3-9-16(10-4-14)19(23,24)25/h1-12H/b11-5+,12-6+

InChI key

OIUBVYQZMUKRRF-YDWXAUTNSA-N

Application

This modified dibenzylideneacetone (dba) ligand was recently reported by Yu and coworkers to assist in the oxidative ortho-C-H borylation of arenes under palladium catalysis. The CF3-dba ligand has also been used in several other applications.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xue-ming Li et al.
Organic letters, 13(3), 374-377 (2010-12-30)
9-Amino-9-deoxyepiquinine efficiently catalyzed the double-conjugate addition of malononitrile to dienones. A number of 1,1,2,6-tetrasubstituted cyclohexanones were prepared in good yields, diastereoselectivities, and excellent enantioselectivities.
Highly Enantioselective Synthesis of Spiro[cyclohexanone-oxindoles] and Spiro[cyclohexanone-pyrazolones] by Asymmetric Cascade [5+1] Double Michael Reactions
Bin Wu, Jian Chen, Mei-Qiu Li, Jin-Xin Zhang, et al.
European Journal of Organic Chemistry, 7, 1318-1327 (2012)
Xin Li et al.
Chemistry, an Asian journal, 8(5), 997-1003 (2013-02-21)
A highly enantioselective catalytic double-Michael addition reaction of substituted benzofuran-2-ones with divinyl ketones promoted by readily accessible tertiary amine-thiourea Cinchona alkaloids has been developed. A number of optically enriched spirocyclic benzofuran-2-ones were prepared in very good yields (up to 99

Related Content

Yu program focuses on efficient C–H bond activation for drug synthesis, using simple starting materials.

Questions

  1. Can you tell us something about the procedure you follow to make this compound?

    1 answer
    1. This product is an M-Clarity level 100 item. The method of preparation is considered proprietary.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service