Skip to Content
Merck
All Photos(2)

Key Documents

ALD00348

Sigma-Aldrich

Boc-3-[3,4-bis(trifluoromethyl)phenyl]-L-alanine

≥95% (HPLC)

Synonym(s):

(S)-3-(3,4-Bis(trifluoromethyl)phenyl)-2-(tert-butoxycarbonylamino)propanoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€117.00

€117.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€117.00

About This Item

Empirical Formula (Hill Notation):
C16H17F6NO4
CAS Number:
Molecular Weight:
401.30
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

€117.00


Check Cart for Availability

Request a Bulk Order

description

ALPH: +24.0, 0.5%EtOH

Quality Level

Assay

≥95% (HPLC)

form

powder or solid

reaction suitability

reaction type: Boc solid-phase peptide synthesis
reagent type: ligand
reaction type: C-H Activation

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

OC([C@@H](NC(OC(C)(C)C)=O)CC1=CC(C(F)(F)F)=C(C(F)(F)F)C=C1)=O

InChI

1S/C16H17F6NO4/c1-14(2,3)27-13(26)23-11(12(24)25)7-8-4-5-9(15(17,18)19)10(6-8)16(20,21)22/h4-6,11H,7H2,1-3H3,(H,23,26)(H,24,25)/t11-/m0/s1

InChI key

LLRXWQAYIGATCH-NSHDSACASA-N

Application

Unnatural amino acid was derived from a C-H Activation methodology developed by Jin-Quan Yu and coworkers. The Pd-mediated C-C bond formation can be made in tandem with 2-Methylpyridine (Aldrich 109835).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service